accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OONHNH2HCl

CAS RN 18807-71-1

IUPAC Name: benzyl N-(2-aminoethyl)carbamate hydrochloride
Synonyms: benzyl N-(2-aminoethyl)carbamate hydrochloride
Molecular Weight (g/mol): 230.69
Molecular Formula: C10H15ClN2O2
InChi Key: QMLKQXIAPAAIEJ-UHFFFAOYSA-N
SMILES: Cl.NCCNC(=O)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results