accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1882-69-5

IUPAC Name: 5-methoxy-2-nitrobenzoate
Synonyms: 5-methoxy-2-nitrobenzoate
Molecular Weight (g/mol): 196.14
Molecular Formula: C8H6NO5
InChi Key: URADKXVAIGMTEG-UHFFFAOYSA-M
SMILES: COC1=CC=C(C(=C1)C([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results