accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3O(E)

CAS RN 18829-55-5

IUPAC Name: (2E)-hept-2-enal
Synonyms: 2-heptenal, (2E)-
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: NDFKTBCGKNOHPJ-AATRIKPKSA-N
SMILES: CCCC\C=C\C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results