CAS RN 188399-46-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
ent-Entecavir, TRC
CAS: 188399-46-4 Molecular Formula: C12H15N5O3 Synonym: 2-Amino-1,9-dihydro-9-[(1R,3S,4R)-4-hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl]-6H-Purin-6-one∣[1R-(1α,3α,4β)]-2-Amino-1,9-dihydro-9-[4-hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl]-6H-purin-6-one;