accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 18856-68-3

IUPAC Name: 1,3-diethyl 2-[(dimethylamino)methylidene]propanedioate
Synonyms: 1,3-diethyl 2-[(dimethylamino)methylidene]propanedioate
Molecular Weight (g/mol): 215.25
Molecular Formula: C10H17NO4
InChi Key: XNSXXTKUKYWLAH-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=CN(C)C)C(=O)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results