accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 188614-01-9

IUPAC Name: methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate
Synonyms: methyl 3-oxo-2,4-dihydro-1,4-benzothiazine-6-carboxylate
Molecular Weight (g/mol): 223.25
Molecular Formula: C10H9NO3S
InChi Key: UFSVRHNRNVVBJR-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC2=C(SCC(=O)N2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results