accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COONNH3CCl

CAS RN 188781-08-0

IUPAC Name: ethyl 2-chloro-4-methylpyrimidine-5-carboxylate
Synonyms: ethyl 2-chloro-4-methylpyrimidine-5-carboxylate
Molecular Weight (g/mol): 200.62
Molecular Formula: C8H9ClN2O2
InChi Key: XJNJPQSXRUJCIB-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C(Cl)N=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results