accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONO

CAS RN 1888-91-1

IUPAC Name: 1-acetylazepan-2-one
Synonyms: 1-acetylazepan-2-one
Molecular Weight (g/mol): 155.2
Molecular Formula: C8H13NO2
InChi Key: QISSLHPKTCLLDL-UHFFFAOYSA-N
SMILES: CC(=O)N1CCCCCC1=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (5)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)