accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3(R)OH(S)(S)H3CH3C

CAS RN 18881-04-4

IUPAC Name: (1S,2R,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-en-2-ol
Synonyms: (1S,2R,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-en-2-ol
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: WONIGEXYPVIKFS-DJLDLDEBSA-N
SMILES: CC1=C[C@@H](O)[C@H]2C[C@@H]1C2(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results