accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOBrI

CAS RN 188815-32-9

IUPAC Name: 3-bromo-5-iodobenzoate
Synonyms: 3-bromo-5-iodobenzoate
Molecular Weight (g/mol): 325.91
Molecular Formula: C7H3BrIO2
InChi Key: MKJBJYCBKXPQSY-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(Br)=CC(I)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results