accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 188837-53-8

IUPAC Name: 2,3,4,5,6-pentafluorophenyl pyridine-2-carboxylate
Synonyms: 2,3,4,5,6-pentafluorophenyl pyridine-2-carboxylate
Molecular Weight (g/mol): 289.16
Molecular Formula: C12H4F5NO2
InChi Key: KFJSLIBBYMXLTG-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=CC=CC=N2)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results