accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3H3C

CAS RN 1889-67-4

IUPAC Name: (2,3-dimethyl-3-phenylbutan-2-yl)benzene
Synonyms: (2,3-dimethyl-3-phenylbutan-2-yl)benzene
Molecular Weight (g/mol): 238.37
Molecular Formula: C18H22
InChi Key: HGTUJZTUQFXBIH-UHFFFAOYSA-N
SMILES: CC(C)(C1=CC=CC=C1)C(C)(C)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results