accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 18902-42-6

RuNOClClCl

CAS RN 18902-42-6

IUPAC Name: trichloro(nitroso)ruthenium
Synonyms: trichloro(nitroso)ruthenium
Molecular Weight (g/mol): 237.43
Molecular Formula: Cl3NORu
InChi Key: AUWSZCCJRSCQEP-UHFFFAOYSA-K
SMILES: Cl[Ru](Cl)(Cl)N=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results