accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOOOOF

CAS RN 189028-93-1

IUPAC Name: 1-(4-fluorophenyl)-5-(2-oxo-4-phenyl-1,3-oxazolidin-3-yl)pentane-1,5-dione
Synonyms: 1-(4-fluorophenyl)-5-(2-oxo-4-phenyl-1,3-oxazolidin-3-yl)pentane-1,5-dione
Molecular Weight (g/mol): 355.37
Molecular Formula: C20H18FNO4
InChi Key: XXSSRSVXDNUAQX-UHFFFAOYNA-N
SMILES: FC1=CC=C(C=C1)C(=O)CCCC(=O)N1C(COC1=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results