accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNC(?)NNCH3CH3

CAS RN 1892-57-5

IUPAC Name: ({[3-(dimethylamino)propyl]imino}methylidene)(ethyl)amine
Synonyms: ({[3-(dimethylamino)propyl]imino}methylidene)(ethyl)amine
Molecular Weight (g/mol): 155.25
Molecular Formula: C8H17N3
InChi Key: LMDZBCPBFSXMTL-UHFFFAOYSA-N
SMILES: CCN=C=NCCCN(C)C
Molecular Weight (g/mol) 
  • (9)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (6)
  • (3)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (9)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)