accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOHOFF

CAS RN 189283-51-0

IUPAC Name: 3,4-difluoro-2-hydroxybenzoic acid
Synonyms: 3,4-difluoro-2-hydroxybenzoic acid
Molecular Weight (g/mol): 174.1
Molecular Formula: C7H4F2O3
InChi Key: GWOOBUWKTOCYKY-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(O)C(F)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results