accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OHNHOOOH3CH3CCH3

CAS RN 189337-28-8

IUPAC Name: (9H-fluoren-9-yl)methyl N-[3-(tert-butoxy)-1-hydroxybutan-2-yl]carbamate
Synonyms: 9H-fluoren-9-ylmethyl N-[3-(tert-butoxy)-1-hydroxybutan-2-yl]carbamate
Molecular Weight (g/mol): 383.49
Molecular Formula: C23H29NO4
InChi Key: LBVPBNDGSCZOTB-UHFFFAOYNA-N
SMILES: CC(OC(C)(C)C)C(CO)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)