accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 18942-49-9

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoate
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3-phenylpropanoate
Molecular Weight (g/mol): 264.3
Molecular Formula: C14H18NO4
InChi Key: ZYJPUMXJBDHSIF-LLVKDONJSA-M
SMILES: CC(C)(C)OC(=O)N[C@H](CC1=CC=CC=C1)C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)