accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 18942-50-2

CH3CH3H3COOHONHOOCH3CH3H3CHN

CAS RN 18942-50-2

IUPAC Name: 3-(tert-butoxy)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid; N-cyclohexylcyclohexanamine
Synonyms: 3-(tert-butoxy)-2-[(tert-butoxycarbonyl)amino]propanoic acid; dicha
Molecular Weight (g/mol): 442.64
Molecular Formula: C24H46N2O5
InChi Key: AIEUUHIXSUNTGV-UHFFFAOYNA-N
SMILES: C1CCC(CC1)NC1CCCCC1.CC(C)(C)OCC(NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)