CAS RN 1899-94-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Methylbenzenesulfonamide, TRC
CAS: 1899-94-1 Molecular Formula: C7H9NO2S Molecular Weight (g/mol): 171.22 IUPAC Name: 3-methylbenzenesulfonamide SMILES: Cc1cccc(c1)S(=O)(=O)N