accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 19004-19-4

H2OH2OH2OH2OH2OCuCuONOOONOOONOOONOO

CAS RN 19004-19-4

IUPAC Name: dicopper(2+) pentahydrate tetranitrate
Synonyms: dicopper(2+) pentahydrate tetranitrate
Molecular Weight (g/mol): 465.18
Molecular Formula: Cu2H10N4O17
InChi Key: IZDJJEMZQZQQQQ-UHFFFAOYSA-N
SMILES: O.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)
Percent Purity 
  • (7)
  • (4)
  • (3)
Grade 
  • (4)
  • (6)
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (1)
Physical Form 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (8)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)
  • (7)
  • (4)
  • (3)
  • (4)
  • (6)
  • (3)
  • (3)
  • (1)
  • (4)
  • (3)