accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1904-24-1

H3CNHNNH2

CAS RN 1904-24-1

IUPAC Name: 5-ethyl-1H-pyrazol-3-amine
Synonyms: 5-ethyl-1H-pyrazol-3-amine
Molecular Weight (g/mol): 111.15
Molecular Formula: C5H9N3
InChi Key: AXDGPQLEVYSXNL-UHFFFAOYSA-N
SMILES: CCC1=CC(N)=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)