accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3SClOO

CAS RN 19040-62-1

IUPAC Name: 2,5-dimethylbenzene-1-sulfonyl chloride
Synonyms: 2,5-dimethylbenzenesulfonyl chloride
Molecular Weight (g/mol): 204.67
Molecular Formula: C8H9ClO2S
InChi Key: FZVZUIBYLZZOEW-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(=C1)S(Cl)(=O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)