accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

KKHO(R)O(R)OPOOO(R)OH(S)HO(R)HO

CAS RN 19046-60-7

IUPAC Name: dipotassium (2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl phosphate
Synonyms: dipotassium α-D-galactose 1-phosphate(2-)
Molecular Weight (g/mol): 336.32
Molecular Formula: C6H11K2O9P
InChi Key: KCIDZIIHRGYJAE-YGFYJFDDSA-L
SMILES: [K+].[K+].OC[C@H]1O[C@H](OP([O-])([O-])=O)[C@H](O)[C@@H](O)[C@H]1O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)