CAS RN 190661-29-1
IUPAC Name:
[2-(benzyloxy)phenyl]boronic acid
Synonyms:
2-(benzyloxy)phenylboronic acid
Molecular Weight (g/mol):
228.05
Molecular Formula:
C13H13BO3
InChi Key:
MCAIDINWZOCYQK-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC=CC=C1OCC1=CC=CC=C1
1
–
2
of
2
results
1
2-Benzyloxybenzeneboronic acid, 96%
CAS: 190661-29-1 Molecular Formula: C13H13BO3 Molecular Weight (g/mol): 228.054 MDL Number: MFCD01632206 InChI Key: MCAIDINWZOCYQK-UHFFFAOYSA-N Synonym: 2-benzyloxyphenylboronic acid,2-benzyloxy phenylboronic acid,2-benzyloxybenzeneboronic acid,2-benzyloxy phenyl boronic acid,2-benzyloxyphenyl boronic acid,2-benzyloxyphenylborornic acid,2-benzyloxy phenyl boranediol,o-benzyloxyphenylboronic acid,boronic acid, 2-phenylmethoxy phenyl PubChem CID: 2773253 IUPAC Name: (2-phenylmethoxyphenyl)boronic acid SMILES: B(C1=CC=CC=C1OCC2=CC=CC=C2)(O)O