accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3OBOCH3CH3N

CAS RN 190788-62-6

IUPAC Name: 8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Synonyms: 8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Molecular Weight (g/mol): 255.12
Molecular Formula: C15H18BNO2
InChi Key: RNTGVJBKPQOGPS-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C2N=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results