accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COFN

CAS RN 191014-55-8

IUPAC Name: 4-fluoro-2-methoxybenzonitrile
Synonyms: 4-fluoro-2-methoxybenzonitrile
Molecular Weight (g/mol): 151.14
Molecular Formula: C8H6FNO
InChi Key: HGBKZVIQHCUHRI-UHFFFAOYSA-N
SMILES: COC1=CC(F)=CC=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results