accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C(S)OH(S)CH3HO

CAS RN 19132-06-0

IUPAC Name: (2S,3S)-butane-2,3-diol
Synonyms: (S,S)-butane-2,3-diol (S,S)-2,3-butylene glycol
Molecular Weight (g/mol): 90.12
Molecular Formula: C4H10O2
InChi Key: OWBTYPJTUOEWEK-IMJSIDKUSA-N
SMILES: C[C@H](O)[C@H](C)O
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)