accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 19155-88-5

IUPAC Name: 2,1,3-benzoxadiazole-5-carboxylate
Synonyms: 2,1,3-benzoxadiazole-5-carboxylate
Molecular Weight (g/mol): 163.11
Molecular Formula: C7H3N2O3
InChi Key: WZUFYJFTOVGJJT-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC2=NON=C2C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results