accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 19168-23-1

NH4NH4ClPdClClClClCl

CAS RN 19168-23-1

IUPAC Name: diammonium hexachloropalladiumdiuide
Synonyms: ammonium hexachloropalladate
Molecular Weight (g/mol): 361.25
Molecular Formula: Cl6H14N2Pd
InChi Key: FOIDYKNKXIWCRM-UHFFFAOYSA-P
SMILES: [NH4+].[NH4+].Cl.Cl.Cl.Cl.Cl.Cl.[Pd--]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results