accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1917-60-8

CH3OOOHO

CAS RN 1917-60-8

IUPAC Name: 5-(methoxymethyl)furan-2-carboxylic acid
Synonyms: 5-(methoxymethyl)furan-2-carboxylic acid
Molecular Weight (g/mol): 156.14
Molecular Formula: C7H8O4
InChi Key: HUUCEXDTXVRYMK-UHFFFAOYSA-N
SMILES: COCC1=CC=C(O1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results