accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SOHO

CAS RN 1918-79-2

IUPAC Name: 5-methylthiophene-2-carboxylic acid
Synonyms: 5-methylthiophene-2-carboxylic acid
Molecular Weight (g/mol): 142.17
Molecular Formula: C6H6O2S
InChi Key: VCNGNQLPFHVODE-UHFFFAOYSA-N
SMILES: CC1=CC=C(S1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)