accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1920-90-7

IUPAC Name: tetrabutylplumbane
Synonyms: tetrabutylplumbane
Molecular Weight (g/mol): 435.7
Molecular Formula: C16H36Pb
InChi Key: KDQHJGWPOQNCMI-UHFFFAOYSA-N
SMILES: CCCC[Pb](CCCC)(CCCC)CCCC
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)