accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 19227-11-3

IUPAC Name: N'-hydroxy-2-phenylethanimidamide
Synonyms: N'-hydroxy-2-phenylethanimidamide
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: FVYBAJYRRIYNBN-UHFFFAOYSA-N
SMILES: NC(CC1=CC=CC=C1)=NO
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)