accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CNCS

CAS RN 19241-16-8

IUPAC Name: 2-isothiocyanato-1,3-dimethylbenzene
Synonyms: 2-isothiocyanato-1,3-dimethylbenzene
Molecular Weight (g/mol): 163.24
Molecular Formula: C9H9NS
InChi Key: UULUECCNPPJFBU-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1N=C=S
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)