accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 192702-79-7

CH3OHF

CAS RN 192702-79-7

IUPAC Name: (3-fluoro-4-methylphenyl)methanol
Synonyms: (3-fluoro-4-methylphenyl)methanol
Molecular Weight (g/mol): 140.16
Molecular Formula: C8H9FO
InChi Key: UUVJAGTYIVYFOS-UHFFFAOYSA-N
SMILES: CC1=CC=C(CO)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results