accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 192725-17-0

H3CH3C(S)NHNOOHN(S)(S)OH(S)HNOOCH3H3C

CAS RN 192725-17-0

IUPAC Name: (2S)-N-[(2S,4S,5S)-5-[2-(2,6-dimethylphenoxy)acetamido]-4-hydroxy-1,6-diphenylhexan-2-yl]-3-methyl-2-(2-oxo-1,3-diazinan-1-yl)butanamide
Synonyms: lopinavir LPV lopinavir (product) aluviran aluvia lopinavir preparation koletra kaletra lopinavir (substance)
Molecular Weight (g/mol): 628.81
Molecular Formula: C37H48N4O5
InChi Key: KJHKTHWMRKYKJE-SUGCFTRWSA-N
SMILES: CC(C)[C@H](N1CCCNC1=O)C(=O)N[C@H](C[C@H](O)[C@H](CC1=CC=CC=C1)NC(=O)COC1=C(C)C=CC=C1C)CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results