accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONOCH3SOOBr

CAS RN 192767-23-0

IUPAC Name: 4-bromo-N,N-bis[(4-methoxyphenyl)methyl]benzene-1-sulfonamide
Synonyms: 4-bromo-N,N-bis[(4-methoxyphenyl)methyl]benzenesulfonamide
Molecular Weight (g/mol): 476.39
Molecular Formula: C22H22BrNO4S
InChi Key: YYLDNQJUMNGLLQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(CN(CC2=CC=C(OC)C=C2)S(=O)(=O)C2=CC=C(Br)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results