accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOBrOH

CAS RN 192810-12-1

IUPAC Name: methyl 3-bromo-5-hydroxybenzoate
Synonyms: methyl 3-bromo-5-hydroxybenzoate
Molecular Weight (g/mol): 231.05
Molecular Formula: C8H7BrO3
InChi Key: KRTBWIIGEJIUSA-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(O)=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results