accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 19285-83-7

IUPAC Name: 2-aminopentanedioic acid hydrate
Synonyms: glutaminsaeure hydrate
Molecular Weight (g/mol): 165.15
Molecular Formula: C5H11NO5
InChi Key: OZDAOHVKBFBBMZ-UHFFFAOYNA-N
SMILES: O.NC(CCC(O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)