accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONNHFONOHOH2OHCl

CAS RN 192927-63-2

IUPAC Name: 1-cyclopropyl-6-fluoro-8-methoxy-7-{octahydro-1H-pyrrolo[3,4-b]pyridin-6-yl}-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrate hydrochloride
Synonyms: moxifloxacin hydrate hydrochloride
Molecular Weight (g/mol): 455.91
Molecular Formula: C21H27ClFN3O5
InChi Key: SKZIMSDWAIZNDD-UHFFFAOYNA-N
SMILES: O.Cl.COC1=C2N(C=C(C(O)=O)C(=O)C2=CC(F)=C1N1CC2CCCNC2C1)C1CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results