accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaNNN

CAS RN 1934-75-4

IUPAC Name: sodium dicyanoazanide
Synonyms: sodium dicyanoazanide
Molecular Weight (g/mol): 89.03
Molecular Formula: C2N3Na
InChi Key: IXBPPZBJIFNGJJ-UHFFFAOYSA-N
SMILES: [Na+].N#C[N-]C#N
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results