accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 193414-60-7

IUPAC Name: 7-bromo-3,4-dihydro-2H-1,4-benzothiazine
Synonyms: 7-bromo-3,4-dihydro-2H-1,4-benzothiazine
Molecular Weight (g/mol): 230.12
Molecular Formula: C8H8BrNS
InChi Key: QWWYGAWGUCQKDL-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(NCCS2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results