accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 19353-92-5

IUPAC Name: 4-(dimethylamino)benzohydrazide
Synonyms: 4-(dimethylamino)benzohydrazide
Molecular Weight (g/mol): 179.22
Molecular Formula: C9H13N3O
InChi Key: HITIGLAGJBMISF-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)C(=O)NN
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results