accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 19381-50-1

NOOHSOOONOOHSOOONOOHSOOONaNaNaFe

CAS RN 19381-50-1

IUPAC Name: trisodium tris(6-hydroxy-5-nitrosonaphthalene-2-sulfonate) iron
Synonyms: (acid green 1
Molecular Weight (g/mol): 881.47
Molecular Formula: C30H18FeN3Na3O15S3
InChi Key: VLBLPLXOYXEXJK-UHFFFAOYSA-K
SMILES: [Na+].[Na+].[Na+].[Fe].OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O.OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O.OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O
Quantity 
  • (2)
  • (2)
  • (1)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)