accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1939-99-7

ClSOO

CAS RN 1939-99-7

IUPAC Name: phenylmethanesulfonyl chloride
Synonyms: phenylmethanesulfonyl chloride
Molecular Weight (g/mol): 190.64
Molecular Formula: C7H7ClO2S
InChi Key: OAHKWDDSKCRNFE-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (3)
Boiling Point 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)