accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 193954-28-8

IUPAC Name: (3S)-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-phenylbutanoic acid
Synonyms: (3S)-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-phenylbutanoic acid
Molecular Weight (g/mol): 401.46
Molecular Formula: C25H23NO4
InChi Key: DQNUGHJJKNFCND-SFHVURJKSA-N
SMILES: OC(=O)C[C@H](CC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results