accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClHSnHClCH3CH3H3CCH3CH3H3C

CAS RN 19429-30-2

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 305.86
Molecular Formula: C8H20Cl2Sn
InChi Key: SCSKBYFEPDHNQR-UHFFFAOYSA-N
SMILES: Cl.Cl.CC(C)(C)[SnH2]C(C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)