accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OSOOSOOOSOOONaNaNa

CAS RN 19437-42-4

IUPAC Name: sodium 3,6-disulfonaphthalene-1-sulfonate
Synonyms: sodium 3,6-disulfonaphthalene-1-sulfonate
Molecular Weight (g/mol): 390.33
Molecular Formula: C10H7NaO9S3
InChi Key: BERQWQSQOIEUMA-UHFFFAOYSA-M
SMILES: [Na+].OS(=O)(=O)C1=CC=C2C(=C1)C=C(C=C2S([O-])(=O)=O)S(O)(=O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results