accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 19438-60-9

IUPAC Name: 5-methyl-octahydro-2-benzofuran-1,3-dione
Synonyms: methylhexahydrophthalic anhydride
Molecular Weight (g/mol): 168.19
Molecular Formula: C9H12O3
InChi Key: FKBMTBAXDISZGN-UHFFFAOYNA-N
SMILES: CC1CCC2C(C1)C(=O)OC2=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)